Ovation M
Actual product may vary
Fisherbrand
Ovation M
The Ovation M is the ideal ergonomic, manual pipette that is autoclavable for easy sterilization.
Have Questions?
Catalog NumberVolume (Metric)
FKS1010112120 to 200 μL
Catalog number FKS10101121
Price (MYR)
6,445.00
Pack
Volume (Metric):
20 to 200 μL
Price (MYR)
6,445.00
Pack
  • Mechanical 2-stroke (overblow) low-force plunger operation
  • Mechanical precise click stop volume adjustment
  • Autoclavable for easy sterilization
  • Built-in microtube cap opener
  • Shelf hanger included with each pipette
  • Two-year warranty
Specifications
Accuracy±0.8% at 200 μL, ±0.8% at 10 μL, ±2.5% (at 20 μL)
AutoclavableAutoclavable
ColorBlue
DisposableReusable
For Use WithPipette Tips
Grip TypeErgonomic
MaterialPolycarbonate ABS
Number of Channels1
Product LineOvation M
ProgrammabilityNo
SterilityNon-sterile
TypeManual Pipette
Volume (Metric)20 to 200 μL
IncludesMicrotube Cap Opener
Increments0.2 μL
Precision±0.15% at 200 μL, ±0.25% at 100 μL, ±1% (at 20 μL)
Quantity1 Ea.
Warranty2 Years
Length (English)10 in.
Width (English)7 in.
Height (English)3 in.
Unit SizePack