Whatman™ Grade 597 Qualitative Filter Circles
Whatman™ Grade 597 Qualitative Filter Circles
Actual product may vary
Whatman™ Grade 597 Qualitative Filter Circles
Filters carbon dioxide and turbidity during beverage analysis. Whatman™ Grade 597 Qualitative Filter Circles may also be used to determine fat content in food analysis.
Have Questions?
Catalog NumberDiameter (Metric)
FKS10098080150 mm
Catalog number FKS10098080
Price (MYR)
387.00
PK
Add to cart
Diameter (Metric):
150 mm
Price (MYR)
387.00
PK
Add to cart
Filters carbon dioxide and turbidity during beverage analysis. Whatman™ Grade 597 Qualitative Filter Circles may also be used to determine fat content in food analysis.
Cellulose filters used in food testing and for the removal of CO2 and turbidity from beverages.
  • Thin filter paper
  • Medium-fast
  • Medium to fine particle retention
Specifications
DescriptionGrade 597 Qualitative Filter Paper Standard Grade, circle, 150 mm
FormatDisc
MaterialCellulose
Particle Retention7 μm
Pore Size4 to 7 μm
Filtration SpeedApprox. 140 s
Flow RateMedium Fast
For Use With (Application)A standard grade filter used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).
GradeGrade 597
Quantity100 Pk.
Thickness (Metric)180 μm
Diameter (Metric)150 mm
Unit SizePK