SuperBlock™ Blocking Buffer
SuperBlock™ Blocking Buffer
Actual product may vary
SuperBlock™ Blocking Buffer
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.
Have Questions?
Catalog NumberQuantityProduct TypeBuffer
FKS100767131 LBlocking BufferPBS
Catalog number FKS10076713
Price (MYR)
-
Quantity:
1 L
Product Type:
Blocking Buffer
Buffer:
PBS
SuperBlock Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis. Blocking micro-plates, membranes, or tissues with a SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer include:
Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Specifications
Product TypeBlocking Buffer
Quantity1 L
FormLiquid
For Use With (Application)ELISA, Western Blot
Product LineSuperBlock™
Concentration1X
BufferPBS
Blocking AgentSingle purified protein
Unit SizeEA
Contents & Storage
Upon receipt store at 4°C. Product is shipped at ambient temperature.