SenTix™ 82 pH Electrode
SenTix™ 82 pH Electrode
Actual product may vary
SenTix™ 82 pH Electrode
pH electrodes with gel electrolyte are ideal for portable measurement but also for routine measurement in the lab
Have Questions?
Catalog NumberMaterial
FKS10069750Glass Shaft, Platinum Diaphragm
Catalog number FKS10069750
Price (MYR)
4,562.00
EA
Add to cart
Material:
Glass Shaft, Platinum Diaphragm
Price (MYR)
4,562.00
EA
Add to cart
pH electrodes with gel electrolyte are ideal for portable measurement but also for routine measurement in the lab. The electrodes do not require any maintenance besides occasional cleaning and are therefore especially economical. For demanding measurements in the lab, there are SenTix™ electrodes with 3 mol/l KCl liquid electrolyte, easy to clean glass shaft and platinum diagphragm. These electrodes can also be used in difficult samples. And if you need an electrode with liquid electrolyte for portable measuring: The SenTix™ 51/52 with plastic shaft, integrated temperature sensor and ceramic diaphragma masters nearly every measuring task, even in the field.
Specifications
Connector TypesBNC, Banana Plug
JunctionPlatinum
MaterialGlass Shaft, Platinum Diaphragm
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypeSenTix™ pH Electrode
For Use With (Equipment)Vario pH meter
IncludesBNC, banana electrode plug, 1m cable
ModelSenTix™ 82
Probe30kilohms Built-in NTC
Storage RequirementsImmerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature (Metric) Range0°C to 100°C
Length (Metric) Cable1 m
Length (Metric) Stem120 mm
Unit SizeEA