Eutech™ ECFG7350401B pH electrode
Product Image
Fisherbrand
Eutech™ ECFG7350401B pH electrode
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
FKS10066474BNC
Catalog number FKS10066474
Price (MYR)
1,485.00
EA
Add to cart
Connector Types:
BNC
Price (MYR)
1,485.00
EA
Add to cart
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This Eutech™ pH electrode is a general purpose glass-body pH electrode with protective sensor guard. Suitable for samples with low temperature measurements. Comes with 10 ml refilling electrolyte is used for general pH measurements. Available with 1 m cable.
Specifications
  • Parameter: pH
  • Range: 0 to 13 pH
  • Temperature Range: 0 to 100 °C
  • Liquid Junction Type: Annular ceramic
  • Internal Reference Type: Ag/AgCl
  • Sealed/Refillable: Refillable
  • Reference Junction: Single
  • Refilling Reference Electrolyte: ECRE001
  • Dimensions (Shaft): 130 x 12mm
  • Cable Length: 1m
  • Connector: BNC
  • Used With: All pHmeters with BNC input connector
  • Specifications
    DescriptionGeneral Purpose Glass-body Single Junction Refillable pH Combination Electrode, annular ceramic reference junction with protective sensor guard, 12x11mm, BNC connector, 1m cable & 1ml refilling electrolyte
    Connector TypesBNC
    JunctionSingle
    MaterialGlass Body
    pH Range0 to 13
    Temperature (Metric) Operating0°C to 100°C
    TypepH Electrode
    Unit SizeEA