Specifications
For Use WithIndustrial Sample Handling of Acids and Alkalis, Not for Heavily Oxidising Acids and Alkalis
MaterialPVDF
Resistance PropertiesChlorine Bleach
Flow Rate20 L/min.
IncludesUniversal bungs made of PVC, for container openings of 30 mm to 70 mm dia.
Unit SizeEA