Nunc™ Centrifuge Bottle Adapters and Cushions
Product Image
Nunc™ Centrifuge Bottle Adapters and Cushions
Increase lab equipment functionality with Thermo Scientific™ Nunc™ Centrifuge Bottle Adapters/Cushions, which are tightness tested, fit most centrifuges and meet USP Class VI.
Have Questions?
Catalog NumberCapacity
FKS10019010200 mL
Catalog number FKS10019010
Price (MYR)
1,798.00
PK
Add to cart
Capacity:
200 mL
Price (MYR)
1,798.00
PK
Add to cart
Increase the functionality of lab equipment with Thermo Scientific™ Nunc™ Centrifuge Bottle Adapters/Cushions, which are tightness tested, fit most centrifuges and meet USP Class VI. Nunc products are high-quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeAdapter
Tube TypeConical
Product LineNunc
MaterialPlastic
Capacity200 mL
For Use With (Equipment)200 mL Tubes
DescriptionCushion Adapter, Nylon, For 200mL tubes
No. per Pack2
Unit SizePK