Mini UV Lamps
Mini UV Lamps
Actual product may vary
Mini UV Lamps
Battery operated and used as portable lamp for industrial, laboratory, lapidary and field inspections.
Have Questions?
Catalog NumberWavelengthLight Source
FKS10015274365 nmUV-A
Catalog number FKS10015274
Price (MYR)
612.00
EA
Add to cart
Wavelength:
365 nm
Light Source:
UV-A
Price (MYR)
612.00
EA
Add to cart
  • Battery-operated 4 watt lamps with lightweight, pocket-size design
  • Features impact-resistant plastic housing with attached nylon strap
  • Select from models with 365nm UV or a combination of 254/365
  • Applications include fluorescence and inspection for field use
Specifications
Light SourceUV-A
Wattage4 W
Wavelength365 nm
Case MaterialPlastic
Depth (English)2.2 in.
Depth (Metric)56 mm
Dimensions (L x W)6.25 x 1 in. (159 x 25 mm)
For Use With (Application)Fluorescence and inspection for field use, industrial inspections, laboratory and lapidary
Length (English)6.25 in.
Length (Metric)159 mm
Packaging Quantity1 Ea.
Width (English)1 in.
Width (Metric)25 mm
Product TypeHandheld Mini UV Lamp
Unit SizeEA