No-Wire™ Cuvet Rack
No-Wire™ Cuvet Rack
Actual product may vary
No-Wire™ Cuvet Rack
No-scratch support for delicate spectrophotometer cells
Have Questions?
Catalog NumberHolds
FKS10003054Light Path Cells (10 mm)
Catalog number FKS10003054
Price (MYR)
440.00
EA
Add to cart
Holds:
Light Path Cells (10 mm)
Price (MYR)
440.00
EA
Add to cart
No-scratch support for delicate spectrophotometer cells
  • Polypropylene rack safely holds light path cuvets in a 6 x 7 array
  • End plates accept labels for quick identification
  • Steam autoclavable at 121°C (250°F)
  • Ships flat; Assembles easily without tools
  • White
  • Stackable
  • Dimensions 5.1L x 4.1W x 1.7 in. H (13 x 11 x 4.3 cm)
Specifications
Product TypeCuvet Rack
AutoclavableAutoclavable
ColorWhite
HoldsLight Path Cells (10 mm)
MaterialPolypropylene
Positions42
ShapeSquare
StackableYes, When Empty
For Use With (Equipment)Cuvet
Dimensions (L x W x H)5 x 4.15 x 1.7 in. (13 x 10.5 x 4.3cm)
Length (Metric)13 cm
Length (English)5 in.
Width (Metric)10.5 cm
Width (English)4.15 in.
Height (Metric)4.3 cm
Height (English)1.7 in.
Unit SizeEA