Kimble™ Wide Tip Opening Reusable Serological Pipets with Plugging Top
Kimble™ Wide Tip Opening Reusable Serological Pipets with Plugging Top
Actual product may vary
Kimble™ Wide Tip Opening Reusable Serological Pipets with Plugging Top
For use with viscous liquids, slurries and suspensions. Wide tip opening for fast delivery.
Have Questions?
Catalog NumberVolume (Metric)Length (Metric)
FKI37034B-2525 mL400 mm
Catalog number FKI37034B-25
Price (MYR)
-
Volume (Metric):
25 mL
Length (Metric):
400 mm
For use with viscous liquids, slurries and suspensions. Wide tip opening for fast delivery.
  • Constricted top for cotton-plugging
  • Color-coded options: 5mL (blue), 10mL (orange), 25mL (white)
  • Tip opening 3mm
  • Graduated: graduation intervals 0.1mL
  • Designed from ASTM Specification E1044, Style III requirements
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
CalibratedYes
Certifications/ComplianceASTM E1044
Diameter (Metric) InnerLarge
MaterialBorosilicate Glass
GraduatedYes
Graduations0 to 24 mL, 0.1 mL increments
Tolerance±0.2 mL
ColorWhite
Volume (Metric)25 mL
Length (Metric)400 mm
TypeSerological Pipet
Unit SizeCS