Check Order Status
Trichloroacetic Acid, for Electrophoresis, Fisher Chemical™
Element Search
Structure Search
Trichloroacetic Acid, for Electrophoresis, Fisher Chemical™
Have Questions?
Change view
Quantity:
250 g
1 kg
Catalog number
FKCTP50050
also known as FKCT/P500/50
Price (MYR)
235.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
235.00
EA
Add to cart
Specifications
pH
1.2
CAS
76-03-9
Melting Point
52°C
Boiling Point
196°C
Color
White
View more