2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
2,3,5-Triphenyltetrazolium, Technical, Fisher Chemical™
C19H15ClN4, CAS Number-298-96-4, 2,3,5-triphenyltetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride, red tetrazolium, tetrazolium red, tetrazolium chloride, uroscreen, urocheck, vitastain, 25g, CHEBI:78019, White, 334.81g/mol | CAS: 298-96-4 | C19H15ClN4 | 334.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FKCT370048
also known as FKCT/3700/48
Price (MYR)
2,968.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,968.00
EA
Add to cart
Specifications
CAS298-96-4
Melting Point235°C
ColorWhite
PackagingPlastic powder jar
Molecular FormulaC19H15ClN4
View more
C19H15ClN4, CAS Number-298-96-4, 2,3,5-triphenyltetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride, red tetrazolium, tetrazolium red, tetrazolium chloride, uroscreen, urocheck, vitastain, 25g, CHEBI:78019, White, 334.81g/mol