Check Order Status
2,2,4-Trimethylpentane 95+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
2,2,4-Trimethylpentane 95+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCT360817
also known as FKCT/3608/17
Price (MYR)
704.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
704.00
EA
Add to cart
Specifications
Melting Point
-107°C
CAS
540-84-1
Color
Colorless
Boiling Point
98°C
Quantity
2.5 L
View more