2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™
2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™
2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™
2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™
2,2,4-Trimethylpentane, for HPLC, Fisher Chemical™
C8H18, CAS Number-540-84-1, 2, 2, 4-trimethylpentane, 2,4,4-trimethylpentane, ch3 2chch2c ch3 3, i-octane, isobutyltrimethylethane, isobutyltrimethylmethane, isooctane, iso-octane, pentane, 2,2,4-trimethyl, unii-qab8f5669o, 2.5L, 98 deg.C, CHEBI:62805, Colorless, 114.23g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Amber Glass Bottle
Catalog number FKCT360615
also known as FKCT/3606/15
Price (MYR)
285.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
285.00
EA
Add to cart
Specifications
Melting Point-107°C
CAS540-84-1
ColorColorless
Boiling Point98°C
Quantity1 L
View more
C8H18, CAS Number-540-84-1, 2, 2, 4-trimethylpentane, 2,4,4-trimethylpentane, ch3 2chch2c ch3 3, i-octane, isobutyltrimethylethane, isobutyltrimethylmethane, isooctane, iso-octane, pentane, 2,2,4-trimethyl, unii-qab8f5669o, 2.5L, 98 deg.C, CHEBI:62805, Colorless, 114.23g/mol