Check Order Status
2,2,4-Trimethylpentane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
2,2,4-Trimethylpentane, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Can
Catalog number
FKCT360125
also known as FKCT/3601/25
Price (MYR)
2,964.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
2,964.00
EA
Add to cart
Specifications
Melting Point
-107°C
CAS
540-84-1
Color
Colorless
Boiling Point
98°C
Quantity
25 L
View more