Check Order Status
Triethylamine, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Triethylamine, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
Amber glass bottle
Catalog number
FKCT320017
also known as FKCT/3200/17
Price (MYR)
415.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
415.00
EA
Add to cart
Specifications
CAS
121-44-8
Melting Point
-115°C
Color
Colorless
pH
12.4
Boiling Point
90°C
View more