Check Order Status
Triethanolamine, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Triethanolamine, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Catalog number
FKCT316017
also known as FKCT/3160/17
Price (MYR)
443.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
443.00
EA
Add to cart
Specifications
CAS
102-71-6
Melting Point
21°C
Color
Yellow
pH
10.5
Boiling Point
360°C
View more