Toluene, Certified AR for Analysis, Fisher Chemical™
Toluene, Certified AR for Analysis, Fisher Chemical™
Toluene, Certified AR for Analysis, Fisher Chemical™
Toluene, Certified AR for Analysis, Fisher Chemical™
Toluene, Certified AR for Analysis, Fisher Chemical™
C7H8, CAS Number-108-88-3, monomethyl benzene, phenylmethane, methylbenzene, methylbenzol, benzene, methyl, methacide, antisal 1a, tolu-sol, toluen, toluol, 2.5L, 111 deg.C, CHEBI:17578, Colorless, 92.14g/mol, YXFVVABEGXRONW-UHFFFAOYSA-N, toluene, -95 deg.C, 8512, 92.141, Amber glass bottle, Liquid
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 L
Packaging:
View all
Plastic-coated Glass Bottle
Amber Glass Bottle
Catalog number FKCT2300PB17
also known as FKCT/2300/PB17
Price (MYR)
183.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
183.00
EA
Add to cart
Specifications
Melting Point-95°C
CAS108-88-3
ColorColorless
Boiling Point111°C
Quantity2.5 L
View more
C7H8, CAS Number-108-88-3, monomethyl benzene, phenylmethane, methylbenzene, methylbenzol, benzene, methyl, methacide, antisal 1a, tolu-sol, toluen, toluol, 2.5L, 111 deg.C, CHEBI:17578, Colorless, 92.14g/mol, YXFVVABEGXRONW-UHFFFAOYSA-N, toluene, -95 deg.C, 8512, 92.141, Amber glass bottle, Liquid