Check Order Status
Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
250 mL
100 mL
Catalog number
FKCT1000PB07
also known as FKCT/1000/PB07
Price (MYR)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
CAS
68-11-1
Melting Point
-16°C
Color
Colorless
pH
1.5
Boiling Point
220°C
View more
Thioglycolic Acid, Extra Pure, SLR