(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number FKCT025060
also known as FKCT/0250/60
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
CAS87-69-4
Melting Point168°C
ColorWhite
pH1.6
PackagingPlastic powder jar
View more
(+)-Tartaric Acid, Certified AR for Analysis, meets analytical specification of Ph.Eur