Check Order Status
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Fisher Chemical™
Element Search
Structure Search
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
2.5 L
Packaging:
HDPE plastic bottle
Catalog number
FKCS9160PB15
also known as FKCS/9160/PB15
Price (MYR)
136.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
136.00
EA
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-93-9
Melting Point
10°C
Boiling Point
290°C
View more