Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Actual product may vary
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Sulfamic Acid, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FKCS864153
also known as FKCS/8641/53
500 gPlastic powder jar
Catalog number FKCS864153
also known as FKCS/8641/53
Price (MYR)
159.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Price (MYR)
159.00
BOT
Add to cart
500g, Plastic powder jar
Specifications
CAS5329-14-6
Melting Point200°C
ColorWhite
pH1.2
PackagingPlastic powder jar
Molecular FormulaH3NO3S
MDL Number11603
Quantity500 g
Synonymamidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
InChI KeyIIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILESNS(=O)(=O)O
IUPAC Namesulfamic acid
Molecular Weight (g/mol)97.088
PubChem CID5987
ChEBICHEBI:9330
Formula Weight97.09g/mol
Physical FormSolid
Unit SizeBOT