Check Order Status
Sulfuric Acid, Pure, 50% (w/v), for formaldehyde Determination, Fisher Chemical
Element Search
Structure Search
Sulfuric Acid, Pure, 50% (w/v), for formaldehyde Determination, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FKCS8350PB17
also known as FKCS/8350/PB17
2.5 L
HDPE plastic bottle
Catalog number
FKCS8350PB17
also known as FKCS/8350/PB17
Price (MYR)
252.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
252.00
EA
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-93-9
Boiling Point
120°C
Packaging
HDPE plastic bottle
View more