Check Order Status
Sodium Hydroxide, Certified AR for Analysis, 30% Solution, d=1.33, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide, Certified AR for Analysis, 30% Solution, d=1.33, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
Catalog number
FKCS4950PB15
also known as FKCS/4950/PB15
Price (MYR)
115.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
115.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
Boiling Point
120°C
CAS
1310-73-2
Melting Point
8°C
View more