BOLA™ Antistatic Explosion Proof Tubing Rolls
BOLA™ Antistatic Explosion Proof Tubing Rolls
Actual product may vary
BOLA™ Antistatic Explosion Proof Tubing Rolls
Antistatic explosion proof tubing rolls made of PTFE-EX.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterFlexibilityMaximum Pressure
FKCS204060
also known as FKCS/2040/60
8 mm10 mm100 mm bend radius35 bar
FGCS204060
also known as FGCS/2040/60
8 mm10 mm100 mm bend radius35 bar
Catalog number FKCS204060
also known as FKCS/2040/60
Price (MYR)
340.00
EA
Add to cart
Diameter (Metric) Inner:
8 mm
Diameter (Metric) Outer:
10 mm
Flexibility:
100 mm bend radius
Maximum Pressure:
35 bar
Price (MYR)
340.00
EA
Add to cart
Very good electric conductivity due to a special 'antistatic compound' made of pure PTFE and the finest, highly pure carbon dust (less than 2.5%).
  • Extensive chemical resistance due to PTFE parts
  • Resistance of less than 1000 kOhm according to DIN EN 12115 directive
  • Ideal for light-sensitive substances
Specifications
Diameter (English) Inner0.31 in.
Diameter (Metric) Inner8 mm
MaterialPTFE
Diameter (English) Outer0.39 in.
Diameter (Metric) Outer10 mm
Overall Length (English)16.4 ft.
Overall Length (Metric)5 m
Thickness (English)0.4 in.
Thickness (Metric)1 m
ColorBlack
Flexibility100 mm bend radius
Maximum Pressure35 bar
Unit SizeEA