Check Order Status
Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal, Fisher Chemical™
Element Search
Structure Search
Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal, Fisher Chemical™
CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol
Have Questions?
Change view
Quantity:
500 g
2.5 kg
Packaging:
Plastic powder jar
Catalog number
FKCS204053
also known as FKCS/2040/53
Price (MYR)
198.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
198.00
EA
Add to cart
Specifications
CAS
6131-90-4
Melting Point
58°C
Color
White
pH
8 to 9.5
Packaging
Plastic powder jar
View more
Sodium Acetate Trihydrate, Certified AR for Analysis, Crystal