Check Order Status
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Element Search
Structure Search
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
500 g
5 kg
Packaging:
View all
Plastic Powder Jar
Catalog number
FKCS076160
also known as FKCS/0761/60
Price (MYR)
198.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic Powder Jar
Request bulk or custom format
Price (MYR)
198.00
BOT
Add to cart
Specifications
Color
Brown-Orange
Physical Form
Solid
pH
1.5 to 5
CAS
7631-86-9
Quantity
1 kg
View more
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent