Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
FKCS069753 also known as FKCS/0697/53 | 500 g |
FGCS069748 also known as FGCS/0697/48 | 100 g |
FGCS069750 also known as FGCS/0697/50 | 250 g |
FGCS069753 also known as FGCS/0697/53 | 500 g |