Check Order Status
Salicylic Acid, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Salicylic Acid, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
250 g
500 g
Catalog number
FKCS028060
also known as FKCS/0280/60
Price (MYR)
678.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
678.00
EA
Add to cart
Specifications
CAS
69-72-7
Melting Point
158°C
Color
White
pH
3.6
Boiling Point
211°C
View more