Check Order Status
Resorcinol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Resorcinol, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
250 g
100 g
Catalog number
FKCR020050
also known as FKCR/0200/50
Price (MYR)
368.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
368.00
EA
Add to cart
Specifications
CAS
108-46-3
Melting Point
109°C
Color
Beige
pH
4.4
Boiling Point
281°C
View more