Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
25 L
Packaging:
View all
Plastic-coated Glass Bottle
Catalog number FKCP7960PB17
also known as FKCP/7960/PB17
Price (MYR)
613.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
613.00
EA
Add to cart
Specifications
pH8.5
Melting Point-42°C
CAS110-86-1
ColorColorless
Boiling Point115°C
View more
C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102