Check Order Status
Pyridine, Anhydrous, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Pyridine, Anhydrous, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Catalog number
FKCP7940PB08
also known as FKCP/7940/PB08
Price (MYR)
314.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
314.00
EA
Add to cart
Specifications
CAS
110-86-1
Melting Point
-42°C
Color
Colorless
pH
8.5
Boiling Point
115°C
View more