Check Order Status
Iso-Propyl Acetate, Pure, Fisher Chemical™
Element Search
Structure Search
Iso-Propyl Acetate, Pure, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
Amber glass bottle
Catalog number
FKCP764017
also known as FKCP/7640/17
Price (MYR)
306.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
306.00
EA
Add to cart
Specifications
CAS
108-21-4
Melting Point
-73°C
Color
Colorless
Boiling Point
88.8°C
Packaging
Amber glass bottle
View more