Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
Propan-1-ol, Certified AR for Analysis, Fisher Chemical™
C3H8O, CAS Number-71-23-8, 1-hydroxypropane, optal, propanol, osmosol extra, n-propyl alcohol, n-propanol, ethylcarbinol, 1-propanol, propyl alcohol, propylic alcohol, 2.5L, 97 deg.C, CHEBI:28831, Colorless, 60.1g/mol, BDERNNFJNOPAEC-UHFFFAOYSA-N, propan-1-ol, -127 deg.C, 2941, 60.096
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number FKCP748117
also known as FKCP/7481/17
Price (MYR)
277.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
277.00
BOT
Add to cart
Specifications
pH7
Melting Point-127°C
CAS71-23-8
ColorColorless
Boiling Point97°C
View more
C3H8O, CAS Number-71-23-8, 1-hydroxypropane, optal, propanol, osmosol extra, n-propyl alcohol, n-propanol, ethylcarbinol, 1-propanol, propyl alcohol, propylic alcohol, 2.5L, 97 deg.C, CHEBI:28831, Colorless, 60.1g/mol, BDERNNFJNOPAEC-UHFFFAOYSA-N, propan-1-ol, -127 deg.C, 2941, 60.096