Check Order Status
Propan-1-ol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Propan-1-ol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Packaging:
Amber Glass Bottle
Catalog number
FKCP748008
also known as FKCP/7480/08
Price (MYR)
122.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
122.00
EA
Add to cart
Specifications
pH
7
Melting Point
-127°C
CAS
71-23-8
Color
Colorless
Boiling Point
97°C
View more