Check Order Status
Potassium Hydroxide, Extra Pure, SLR, Pellets, Fisher Chemical™
Element Search
Structure Search
Potassium Hydroxide, Extra Pure, SLR, Pellets, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
1 kg
2 kg
5 kg
Catalog number
FKCP560053
also known as FKCP/5600/53
Price (MYR)
124.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
124.00
EA
Add to cart
Specifications
pH
13.5
Color
White
Boiling Point
1320°C
CAS
1310-58-3
Melting Point
360°C
View more