Check Order Status
Phosphorus Pentoxide, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Phosphorus Pentoxide, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
Catalog number
FKCP300053
also known as FKCP/3000/53
Price (MYR)
315.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
315.00
BOT
Add to cart
Specifications
Melting Point
340°C
CAS
1314-56-3
Color
White
pH
1
Physical Form
Solid
View more
Phosphorus Pentoxide, Certified AR for Analysis