Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113 | CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
2 kg
Catalog number FKCP236060
also known as FKCP/2360/60
Price (MYR)
309.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
309.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenol Detached Crystals
CAS108-95-2
Melting Point39°C
ColorColorless to White
pH6
View more
C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113