Check Order Status
Phenol Detached Crystals, SLR, Extra Pure, Fisher Chemical™
Element Search
Structure Search
Phenol Detached Crystals, SLR, Extra Pure, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 kg
500 g
Catalog number
FKCP232060
also known as FKCP/2320/60
Price (MYR)
275.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
275.00
EA
Add to cart
Specifications
CAS
108-95-2
Melting Point
39°C
Color
Colorless to White
pH
6
Boiling Point
182°C
View more
Phenol Detached Crystals, SLR, Extra Pure