Check Order Status
1,10-Phenanthroline Ferrous Solution 0.025M, Pure, Fisher Chemical
Element Search
Structure Search
1,10-Phenanthroline Ferrous Solution 0.025M, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FKCP2220PB05
also known as FKCP/2220/PB05
100 mL
Catalog number
FKCP2220PB05
also known as FKCP/2220/PB05
Price (MYR)
163.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
163.00
BOT
Add to cart
Specifications
Packaging
Plastic Coated Glass bottle
CAS
14634-91-4
Physical Form
Liquid
Molecular Formula
C36H24FeN6O4S
MDL Number
MFCD00036428
View more