Check Order Status
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
Element Search
Structure Search
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
High purity solvent for GC and pesticide residue analysis
Have Questions?
Change view
Quantity:
2.5 L
4 L
Packaging:
View all
Amber Glass Bottle
Catalog number
FKCP209217
also known as FKCP/2092/17
Price (MYR)
666.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
666.00
EA
Add to cart
Specifications
CAS
64742-49-0
Color
Colorless
Boiling Point
40°C
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more
Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
Also meets ACS specifications
Intended for research use only