Petroleum Ether 100-120°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
Petroleum Ether 100-120&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 100-120&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 100-120&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 100-120°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
C6H14, CAS Number-64742-49-0, unii-49ib0u6mld, pentane, 2-methyl, iso-hexane, 1,1-dimethylbutane, dimethylpropylmethane, 2-methylpentan, 2-methyl-pentane, 2-methyl pentane, methyl pentane, isohexane, 2.5L, 100 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number FKCP188017
also known as FKCP/1880/17
Price (MYR)
183.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
183.00
EA
Add to cart
Specifications
CAS64742-49-0
ColorColorless
Boiling Point100°C
Quantity2.5 L
PackagingAmber Glass Bottle
View more
C6H14, CAS Number-64742-49-0, unii-49ib0u6mld, pentane, 2-methyl, iso-hexane, 1,1-dimethylbutane, dimethylpropylmethane, 2-methylpentan, 2-methyl-pentane, 2-methyl pentane, methyl pentane, isohexane, 2.5L, 100 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178