Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
Petroleum Ether 40-60&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 40-60&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 40-60&deg;C, Certified AR for Analysis, N-Hexane &lt;2%, Fisher Chemical&trade;
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
2.5 L
Packaging:
View all
Metal Can
Catalog number FKCP176025
also known as FKCP/1760/25
Price (MYR)
1,232.00
DR
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,232.00
DR
Add to cart
Specifications
CAS64742-49-0
ColorColorless
Boiling Point40°C
Quantity25 L
PackagingMetal Can
View more
C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178