Check Order Status
Petroleum Ether 100-120°C, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Petroleum Ether 100-120°C, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber Glass Bottle
Catalog number
FKCP156017
also known as FKCP/1560/17
Price (MYR)
191.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
191.00
BOT
Add to cart
Specifications
CAS
64742-49-0
Color
Colorless
Boiling Point
100°C
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more