Check Order Status
Perchloric Acid 70% d=1.67, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Perchloric Acid 70% d=1.67, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
1 L
Catalog number
FKCP1280PB17
also known as FKCP/1280/PB17
Price (MYR)
1,686.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,686.00
BOT
Add to cart
Specifications
pH
0.1
Color
Colorless
CAS
7601-90-3
Melting Point
-18°C
Boiling Point
203°C
View more