Check Order Status
Perchloric Acid 60% d=1.53, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Perchloric Acid 60% d=1.53, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
Plastic Coated Glass Bottle
Catalog number
FKCP1240PB08
also known as FKCP/1240/PB08
Price (MYR)
532.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Plastic Coated Glass Bottle
Request bulk or custom format
Price (MYR)
532.00
EA
Add to cart
Specifications
pH
0.1
Color
Colorless
CAS
7601-90-3
Melting Point
-18°C
Boiling Point
203°C
View more