Check Order Status
N-Pentane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
N-Pentane, Certified AR for Analysis, Fisher Chemical™
C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber glass bottle
Catalog number
FKCP102117
also known as FKCP/1021/17
Price (MYR)
396.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
396.00
BOT
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
109-66-0
Vapor Pressure
573mbar at 20°C
Viscosity
0.25 mPaS at 20°C
Vapor Density
2.5
View more
C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C