Check Order Status
Pentane, for HPLC, Mixed Isomers, Fisher Chemical™
Element Search
Structure Search
Pentane, for HPLC, Mixed Isomers, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCP100617
also known as FKCP/1006/17
Price (MYR)
533.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
533.00
EA
Add to cart
Specifications
Melting Point
-130°C
CAS
109-66-0
Boiling Point
36°C
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more