Check Order Status
Paraffin Liquid, Pure, Water White, d=0.83-0.86, Fisher Chemical™
Element Search
Structure Search
Paraffin Liquid, Pure, Water White, d=0.83-0.86, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Amber glass bottle
Catalog number
FKCP040017
also known as FKCP/0400/17
Price (MYR)
270.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
270.00
EA
Add to cart
Specifications
CAS
8042-47-5
Melting Point
-9°C
Boiling Point
310°C
Packaging
Amber glass bottle
Molecular Formula
C16H10N2Na2O7S2
View more
Paraffin Liquid, Pure, Water White, d=0.83-0.86