Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™
Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™
Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™
Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™
Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™
H3O4P, CAS Number-7664-38-2, acide phosphorique, acido fosforico, acidum phosphoricum, evits, o-phosphoric acid, orthophosphoric acid, phosphorsaeure, sonac, wc-reiniger, white, 2.5L, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994, HDPE plastic bottle | CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
25 L
Packaging:
View all
Plastic Coated Glass bottle
HDPE plastic bottle
Catalog number FKCO0500PC17
also known as FKCO/0500/PC17
Price (MYR)
452.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
452.00
BOT
Add to cart
Specifications
Boiling Point158°C
Chemical Name or MaterialOrthophosphoric Acid
Melting Point21°C
CAS7664-38-2
pH2
View more
H3O4P, CAS Number-7664-38-2, acide phosphorique, acido fosforico, acidum phosphoricum, evits, o-phosphoric acid, orthophosphoric acid, phosphorsaeure, sonac, wc-reiniger, white, 2.5L, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994, HDPE plastic bottle